3-(Methylamino)phenol 95%

Supplier: Ambeed
Warning

A124885-100MG A124885-250MG A124885-1G
76870-448EA 58.35 USD
76870-448 76870-450 77566-406
3-(Methylamino)phenol 95%
3-(Methylamino)phenol

Precaución: For research use only.

Formula: C₇H₉NO
MW: 123.152 g/mol
MDL Number: MFCD16998628
CAS Number: 14703-69-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR