Dicyclohexyl(2',4',6'-triisopropyl-3,6-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine-(2-(2-aminoethyl)phenyl)palladium(II) chloride 98+%

Supplier: Ambeed
Warning

A124341-1G A124341-250MG A124341-5G A124341-100MG
76870-422EA 55 USD
76870-422 76870-424 76870-426 77670-764
Dicyclohexyl(2',4',6'-triisopropyl-3,6-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine-(2-(2-aminoethyl)phenyl)palladium(II) chloride 98+%
Dicyclohexyl(2',4',6'-triisopropyl-3,6-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine-(2-(2-aminoethyl)phenyl)palladium(II) chloride

Precaución: for Research Use Only

Formula: C₄₃H₆₂ClNO₂PPd
MW: 797.8 g/mol
MDL Number: MFCD12545956
CAS Number: 1148148-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR