1-Methyl-1H-pyrazole-5-carbonitrile 95%

Supplier: Ambeed
Danger

A123857-100MG A123857-250MG
76870-386EA 14.86 USD
76870-386 76870-388
1-Methyl-1H-pyrazole-5-carbonitrile 95%
1-Methyl-1H-pyrazole-5-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₅N₃
MDL Number: MFCD09701708
CAS Number: 66121-72-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR