3-Bromo-4-fluorobenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (3-Bromo-4-fluorophenyl)boronic acid, 3-Bromo-4-fluorophenylboronic acid

A123648-100MG A123648-1G A123648-250MG A123648-5G
76870-366EA 47.75 USD
76870-366 76870-368 76870-370 76870-372
3-Bromo-4-fluorobenzeneboronic acid 98%
3-Bromo-4-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrFO₂
MW: 218.816 g/mol
MDL Number: MFCD13195642
CAS Number: 1092533-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR