(2-Ethoxy-3-fluorophenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-2-ethoxyphenylboronic acid

A123547-1G A123547-250MG
76870-362EA 60.48 USD
76870-362 76870-364
(2-Ethoxy-3-fluorophenyl)boronic acid 97%
(2-Ethoxy-3-fluorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
MW: 183.97 g/mol
MDL Number: MFCD11520053
CAS Number: 1010097-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR