4-Fluoro-2-iodobenzonitrile 95%

Supplier: Ambeed
Warning

A123138-25G A123138-5G A123138-1G
76870-340EA 159.14 USD
76870-340 76870-342 77670-746
4-Fluoro-2-iodobenzonitrile 95%
4-Fluoro-2-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃FIN
MW: 247.0100 g/mol
MDL Number: MFCD09998181
CAS Number: 1031929-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR