((2R,3R,4R)-3-(Benzoyloxy)-4-fluoro-4-methyl-5-oxotetrahydrofuran-2-yl)methyl benzoate 97%

Supplier: Ambeed
Warning

A123043-100G A123043-10G A123043-1G A123043-25G A123043-5G A123043-500G
76870-326EA 65.78 USD
76870-326 76870-328 76870-330 76870-332 76870-334 77571-408
((2R,3R,4R)-3-(Benzoyloxy)-4-fluoro-4-methyl-5-oxotetrahydrofuran-2-yl)methyl benzoate 97%
((2R,3R,4R)-3-(Benzoyloxy)-4-fluoro-4-methyl-5-oxotetrahydrofuran-2-yl)methyl benzoate

Precaución: for Research Use Only

Formula: C₂₀H₁₇FO₆
MW: 372.34 g/mol
MDL Number: MFCD17677380
CAS Number: 874638-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR