2-Bromo-6-methoxybenzonitrile 98%

Supplier: Ambeed
Warning

A122547-10G A122547-250MG A122547-5G A122547-25G
76870-306EA 57 USD
76870-306 76870-308 76870-310 77882-988
2-Bromo-6-methoxybenzonitrile 98%
2-Bromo-6-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆BrNO
MDL Number: MFCD09834771
CAS Number: 1245647-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR