Ethyl-5-phenylthiazole-2-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 5-phenylthiazole-2-carboxylate

A122435-100MG A122435-1G A122435-250MG A122435-5G
76870-298EA 81.69 USD
76870-298 76870-300 76870-302 76870-304
Ethyl-5-phenylthiazole-2-carboxylate 95%
Ethyl-5-phenylthiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₂S
MW: 233.2900 g/mol
MDL Number: MFCD11976519
CAS Number: 58333-72-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR