Omeprazole sulfide 99%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridyl)methyl]thio]benzimidazole

A122371-100G A122371-10G A122371-25G A122371-500G A122371-5G
76870-286EA 59.42 USD
76870-286 76870-288 76870-290 76870-292 76870-294
Omeprazole sulfide 99%
Omeprazole sulfide

Precaución: for Research Use Only

Formula: C₁₇H₁₉N₃O₂S
MW: 329.42 g/mol
MDL Number: MFCD00869021
CAS Number: 73590-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR