4-Hydroxy-3-methoxy-5-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A122358-1G A122358-250MG A122358-25G
76870-282EA 8 USD
76870-282 76870-284 77882-986
4-Hydroxy-3-methoxy-5-nitrobenzoic acid 98%
4-Hydroxy-3-methoxy-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₆
MW: 213.1443 g/mol
MDL Number: MFCD06203146
CAS Number: 15785-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR