2-Bromo-5-(trifluoromethyl)thiophene 95%

Supplier: Ambeed
Danger

A122036-100MG A122036-1G A122036-250MG A122036-5G
76870-258EA 80 USD
76870-258 76870-260 76870-262 76870-264
2-Bromo-5-(trifluoromethyl)thiophene 95%
2-Bromo-5-(trifluoromethyl)thiophene

Precaución: for Research Use Only

Formula: C₅H₂BrF₃S
MDL Number: MFCD08461914
CAS Number: 143469-22-1
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR