2-Cyanobenzeneboronic acid-1,3-propanediol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(1,3,2-Dioxaborinan-2-yl)benzonitrile, 2-Cyanobenzeneboronic acid 1,3-Propanediol ester, 2-Cyanophenylboronic acid 1,3-Propanediol ester

A121504-100G A121504-10G A121504-1G A121504-25G A121504-5G
76870-238EA 229.16 USD
76870-238 76870-240 76870-242 76870-244 76870-246
2-Cyanobenzeneboronic acid-1,3-propanediol ester 98%
2-Cyanobenzeneboronic acid-1,3-propanediol ester

Precaución: for Research Use Only

Formula: C₁₀H₁₀BNO₂
MW: 187.00 g/mol
MDL Number: MFCD04039012
CAS Number: 172732-52-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR