1,3-Dichloro-7-fluoroisoquinoline 95%

Supplier: Ambeed
Warning

A120607-100MG A120607-1G A120607-250MG A120607-5G
76870-200EA 48.81 USD
76870-200 76870-202 76870-204 77566-234
1,3-Dichloro-7-fluoroisoquinoline 95%
1,3-Dichloro-7-fluoroisoquinoline

Precaución: For research use only.

Formula: C₉H₄Cl₂FN
MW: 216.0392 g/mol
MDL Number: MFCD09763685
CAS Number: 941294-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR