(R)-8-(Benzyloxy)-5-(2-bromo-1-hydroxyethyl)quinolin-2(1H)-one 97%

Supplier: Ambeed
Warning

A120553-100MG A120553-1G A120553-250MG A120553-5G
76870-192EA 16.98 USD
76870-192 76870-194 76870-196 76870-198
(R)-8-(Benzyloxy)-5-(2-bromo-1-hydroxyethyl)quinolin-2(1H)-one 97%
(R)-8-(Benzyloxy)-5-(2-bromo-1-hydroxyethyl)quinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₁₈H₁₆BrNO₃
MW: 374.22 g/mol
MDL Number: MFCD20484048
CAS Number: 530084-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR