N,N-Dimethylpiperidin-4-amine hydrochloride 95%

Supplier: Ambeed
Warning

A119999-250MG A119999-5G
76870-158EA 8.49 USD
76870-158 76870-160
N,N-Dimethylpiperidin-4-amine hydrochloride 95%
N,N-Dimethylpiperidin-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₇ClN₂
MDL Number: MFCD03938881
CAS Number: 172281-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR