4-Oxo-3,4-dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester 96%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 4-oxo-3,4-dihydropyridine-1(2H)-carboxylate, 4-Oxo-1,2,3,4-tetrahydropyridine, N-BOC protected

A119001-1G A119001-250MG A119001-5G
76870-134EA 125.19 USD
76870-134 76870-136 77571-122
4-Oxo-3,4-dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester 96%
4-Oxo-3,4-dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO₃
MW: 197.24 g/mol
MDL Number: MFCD08437478
CAS Number: 325486-45-1
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR