Pyrimidin-2-ol 95%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxypyrimidine

A118857-100G A118857-10G A118857-1G A118857-25G A118857-5G
76870-124EA 774.46 USD
76870-124 76870-126 76870-128 76870-130 76870-132
Pyrimidin-2-ol 95%
Pyrimidin-2-ol

Precaución: for Research Use Only

Formula: C4H4N2O
MW: 96.0900 g/mol
MDL Number: MFCD00047358
CAS Number: 51953-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR