tert-Butyl-3-formylbenzoate 96%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 3-formylbenzoate

A118537-100MG A118537-250MG A118537-1G
76870-120EA 47.75 USD
76870-120 76870-122 77566-170
tert-Butyl-3-formylbenzoate 96%
tert-Butyl-3-formylbenzoate

Precaución: For research use only.

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
MDL Number: MFCD02180885
CAS Number: 247186-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR