1-Methyl-1H-indole-2-boronic acid, neopentyl glycol ester 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-methyl-1H-indole
A118364-1G
A118364-250MG
A118364-5G
76870-114EA
178
USD
76870-114
76870-116
76870-118
1-Methyl-1H-indole-2-boronic acid, neopentyl glycol ester 96%
1-Methyl-1H-indole-2-boronic acid, neopentyl glycol ester
Precaución: for Research Use Only
Formula:
C₁₄H₁₈BNO₂ |
MDL Number:
MFCD06797198 CAS Number: 905966-48-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...