3,5-Bis(trifluoromethyl)benzylamine 97%

Supplier: Ambeed
Danger

Synonyms: (3,5-Bis(trifluoromethyl)phenyl)methanamine

A118067-10G A118067-1G A118067-25G
76870-102EA 29 USD
76870-102 76870-104 76870-106
3,5-Bis(trifluoromethyl)benzylamine 97%
3,5-Bis(trifluoromethyl)benzylamine

Precaución: for Research Use Only

Formula: C₉H₇F₆N
MDL Number: MFCD00009909
CAS Number: 85068-29-7
UN: 3259
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR