Ethyl 2-(6-methylpyridin-2-yl)acetate 95%

Supplier: Ambeed
Warning

A117833-10G A117833-1G A117833-250MG A117833-5G
76870-086EA 562.28 USD
76870-086 76870-088 76870-090 76870-092
Ethyl 2-(6-methylpyridin-2-yl)acetate 95%
Ethyl 2-(6-methylpyridin-2-yl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
MDL Number: MFCD10698588
CAS Number: 5552-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR