5-Bromo-4-methoxy-2-nitroaniline 98%

Supplier: Ambeed
Warning

A117762-1G A117762-250MG
76870-082EA 209 USD
76870-082 76870-084
5-Bromo-4-methoxy-2-nitroaniline 98%
5-Bromo-4-methoxy-2-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₃
MW: 247.0461 g/mol
MDL Number: MFCD00031127
CAS Number: 6943-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR