(1R,3S,5R)-2-(tert-Butoxycarbonyl)-2-azabicyclo[3.1.0]hexane-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A117728-100MG A117728-1G A117728-250MG A117728-5G
76870-076EA 21 USD
76870-076 76870-078 76870-080 77882-964
(1R,3S,5R)-2-(tert-Butoxycarbonyl)-2-azabicyclo[3.1.0]hexane-3-carboxylic acid 97%
(1R,3S,5R)-2-(tert-Butoxycarbonyl)-2-azabicyclo[3.1.0]hexane-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₄
MDL Number: MFCD08691405
CAS Number: 197142-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR