(R)-(+)-Tetrahydro-2-furoic acid 98%

Supplier: Ambeed
Danger

A117217-100G A117217-10G A117217-1G A117217-25G A117217-5G
76870-054EA 107.16 USD
76870-054 76870-056 76870-058 76870-060 76870-062
(R)-(+)-Tetrahydro-2-furoic acid 98%
(R)-(+)-Tetrahydro-2-furoic acid

Precaución: for Research Use Only

Formula: C₅H₈O₃
MDL Number: MFCD00211271
CAS Number: 87392-05-0
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR