3-Fluoro-5-nitropyridin-2(1H)-one 98%

Supplier: Ambeed
Warning

A117121-5G A117121-1G
76870-052EA 152.77 USD
76870-052 77670-646
3-Fluoro-5-nitropyridin-2(1H)-one 98%
3-Fluoro-5-nitropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₃FN₂O₃
MW: 158.08 g/mol
MDL Number: MFCD10699675
CAS Number: 1033202-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR