(R)-tert-Butyl 3-carbamoylpiperidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A116706-1G A116706-250MG A116706-5G
76870-028EA 71.09 USD
76870-028 76870-030 77549-324
(R)-tert-Butyl 3-carbamoylpiperidine-1-carboxylate 95%
(R)-tert-Butyl 3-carbamoylpiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₃
MW: 228.28 g/mol
MDL Number: MFCD11977492
CAS Number: 915226-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR