2-Amino-p-cresol 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-4-hydroxytoluene, 1-Amino-2-hydroxy-5-methylbenzene, 2-Amino-4-methylphenol, 2-Hydroxy-5-methylaniline

A116373-100G A116373-10G A116373-25G A116373-500G
76870-012EA 21.22 USD
76870-012 76870-014 76870-016 76870-018
2-Amino-p-cresol 97%
2-Amino-p-cresol

Precaución: for Research Use Only

Formula: C₇H₉NO
MW: 123.15 g/mol
MDL Number: MFCD00007699
CAS Number: 95-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR