N-(4-(1-(2-(6-Methylpyridin-2-yl)ethyl)piperidine-4-carbonyl)phenyl)methanesulfonamide dihydrochloride 98%

Supplier: Ambeed
Warning

A115118-10MG A115118-1MG A115118-25MG A115118-50MG A115118-5MG
76869-962EA 96 USD
76869-962 76869-964 76869-966 76869-968 76869-970
N-(4-(1-(2-(6-Methylpyridin-2-yl)ethyl)piperidine-4-carbonyl)phenyl)methanesulfonamide dihydrochloride 98%
N-(4-(1-(2-(6-Methylpyridin-2-yl)ethyl)piperidine-4-carbonyl)phenyl)methanesulfonamide dihydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₂₉Cl₂N₃O₃S
MDL Number: MFCD01754739
CAS Number: 113559-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR