2-(1H-Imidazol-2-yl)pyrazine 98%

Supplier: Ambeed
Warning

A114689-1G A114689-250MG
76869-940EA 87 USD
76869-940 76869-942
2-(1H-Imidazol-2-yl)pyrazine 98%
2-(1H-Imidazol-2-yl)pyrazine

Precaución: for Research Use Only

Formula: C₇H₆N₄
MDL Number: MFCD08668973
CAS Number: 119165-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR