(S)-Methyl morpholine-3-carboxylate hydrochloride 98%

Supplier: Ambeed
Warning

A114590-1G A114590-250MG
76869-936EA 150.65 USD
76869-936 76869-938
(S)-Methyl morpholine-3-carboxylate hydrochloride 98%
(S)-Methyl morpholine-3-carboxylate hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₃
MW: 181.6174 g/mol
MDL Number: MFCD09910333
CAS Number: 1447972-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR