4-Amino-2-chloro-5-fluorobenzonitrile 95%

Supplier: Ambeed
Warning

A114556-1G A114556-250MG A114556-5G
76869-930EA 55 USD
76869-930 76869-932 76869-934
4-Amino-2-chloro-5-fluorobenzonitrile 95%
4-Amino-2-chloro-5-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄ClFN₂
MDL Number: MFCD11110982
CAS Number: 1228376-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR