2-Chloro-4-(4-fluorophenyl)-5,6,7,8,9,10-hexahydrocycloocta[b]pyridine 98%

Supplier: Ambeed
Warning

A114383-100MG A114383-250MG
76869-920EA 35.01 USD
76869-920 76869-922
2-Chloro-4-(4-fluorophenyl)-5,6,7,8,9,10-hexahydrocycloocta[b]pyridine 98%
2-Chloro-4-(4-fluorophenyl)-5,6,7,8,9,10-hexahydrocycloocta[b]pyridine

Precaución: for Research Use Only

Formula: C₁₇H₁₇ClFN
MW: 289.77 g/mol
MDL Number: MFCD09955376
CAS Number: 132813-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR