2-Bromo-3-butylthiophene 97%

Supplier: Ambeed
Warning

A114106-100G A114106-1G A114106-25G A114106-5G
76869-894EA 260.99 USD
76869-894 76869-896 76869-898 76869-900
2-Bromo-3-butylthiophene 97%
2-Bromo-3-butylthiophene

Precaución: for Research Use Only

Formula: C₈H₁₁BrS
MW: 219.14 g/mol
MDL Number: MFCD16619253
CAS Number: 145543-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR