Bis(4-hydroxyphenyl) sulfone 97%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Sulphonyldiphenol, 4,4'-Dihydroxy diphenyl sulphone, 4-Hydroxyphenyl sulphone, 4,4'-sulphonyldiphenol

A114055-100G A114055-25G A114055-500G
76869-888EA 6.37 USD
76869-888 76869-890 76869-892
Bis(4-hydroxyphenyl) sulfone 97%
Bis(4-hydroxyphenyl)sulphone

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₄S
MW: 250.28 g/mol
MDL Number: MFCD00002350
CAS Number: 80-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR