Methyl 2-bromo-4-(trifluoromethyl)benzoate 95%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-(trifluoromethyl)benzoic acid methyl ester

A113940-100MG A113940-10G A113940-1G A113940-250MG A113940-25G A113940-5G
76869-876EA 8.49 USD
76869-876 76869-878 76869-880 76869-882 76869-884 76869-886
Methyl 2-bromo-4-(trifluoromethyl)benzoate 95%
Methyl 2-bromo-4-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₆BrF₃O₂
MW: 283.042 g/mol
MDL Number: MFCD14697995
CAS Number: 1214334-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR