1-Iodo-3,5-dimethoxybenzene 95%

Supplier: Ambeed
Warning

A113599-1G A113599-250MG A113599-5G A113599-25G
76869-870EA 16 USD
76869-870 76869-872 76869-874 77565-992
1-Iodo-3,5-dimethoxybenzene 95%
1-Iodo-3,5-dimethoxybenzene

Precaución: For research use only.

Formula: C₈H₉IO₂
MDL Number: MFCD08729270
CAS Number: 25245-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR