3-Hydroxymethyl-2-methylbiphenyl 97%

Supplier: Ambeed
Warning

Synonyms: (2-Methyl-[1,1'-biphenyl]-3-yl)methanol, 2-Methylbiphenyl-3-methanol

A113515-1000G A113515-100G A113515-10G A113515-25G A113515-500G A113515-5G
76869-856EA 284.33 USD
76869-856 76869-858 76869-860 76869-862 76869-864 76869-866
3-Hydroxymethyl-2-methylbiphenyl 97%
3-Hydroxymethyl-2-methylbiphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₄O
MW: 198.27 g/mol
MDL Number: MFCD00134200
CAS Number: 76350-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR