HEPPSO sodium salt (β-Hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulfonic acid sodium salt) 97%

Supplier: Ambeed
Warning

Synonyms: Sodium 2-hydroxy-1-(4-(2-hydroxyethyl)piperazin-1-yl)propane-1-sulphonate

A113297-100G A113297-10G A113297-1G A113297-25G
76869-840EA 61 USD
76869-840 76869-842 76869-844 76869-846 76869-848
HEPPSO sodium salt (β-Hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulfonic acid sodium salt) 97%
HEPPSO sodium salt (β-Hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulphonic acid sodium salt)

Precaución: for Research Use Only

Formula: C₉H₁₉N₂NaO₅S
MDL Number: MFCD00070006
CAS Number: 89648-37-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR