(E)-4-(Dimethylamino)but-2-enoic acid xhydrochloride 98%

Supplier: Ambeed
Warning

A113251-100G A113251-10G A113251-1G A113251-25G A113251-5G
76869-830EA 1312.34 USD
76869-830 76869-832 76869-834 76869-836 76869-838
(E)-4-(Dimethylamino)but-2-enoic acid xhydrochloride 98%
(E)-4-(Dimethylamino)but-2-enoic acid xhydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₂ClNO₂
MW: 165.61 g/mol
CAS Number: 1130155-48-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR