b-Amino-3-bromobenzeneethanol 98%

Supplier: Ambeed
Warning

A112533-100MG A112533-1G A112533-250MG A112533-5G
76869-808EA 7 USD
76869-808 76869-810 76869-812 77610-894
b-Amino-3-bromobenzeneethanol 98%
b-Amino-3-bromobenzeneethanol

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
MDL Number: MFCD12827191
CAS Number: 188586-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR