(E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein 98%

Supplier: Ambeed
Danger

Synonyms: (E)-3-[3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl]-propenal, (E)-3-(3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl)acrylaldehyde, (E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]propenal

A112502-10G A112502-1G A112502-5G
76869-802EA 19.1 USD
76869-802 76869-804 76869-806
(E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein 98%
(E)-3-[3-(4-Fluorophenyl)-1-isopropylindol-2-yl]acrolein

Precaución: for Research Use Only

Formula: C₂₀H₁₈FNO
MW: 307.37 g/mol
MDL Number: MFCD03840863
CAS Number: 93957-50-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR