N-(3-Acetyl-2-hydroxyphenyl)acetamide 95%

Supplier: Ambeed
Warning

A111964-1G A111964-250MG A111964-5G
76869-792EA 6.37 USD
76869-792 76869-794 76869-796
N-(3-Acetyl-2-hydroxyphenyl)acetamide 95%
N-(3-Acetyl-2-hydroxyphenyl)acetamide

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₃
MW: 193.2 g/mol
MDL Number: MFCD17170001
CAS Number: 103205-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR