Fluconazole 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2,4-Difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol, 2-(2,4-Difluorophenyl)-1,3-di(1H-1,2,4-triazol-1-yl)propan-2-ol

A111824-10G A111824-25G A111824-500G A111824-5G A111824-100G
76869-774EA 19 USD
76869-774 76869-776 76869-778 76869-780 77882-935
Fluconazole 98%
Fluconazole

Precaución: for Research Use Only

Formula: C₁₃H₁₂F₂N₆O
MDL Number: MFCD00274549
CAS Number: 86386-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR