3-(3-Bromophenyl)-4H-1,2,4-triazole 98%

Supplier: Ambeed
Warning

A111778-100MG A111778-1G
76869-772EA 50.93 USD
76869-772 77549-170
3-(3-Bromophenyl)-4H-1,2,4-triazole 98%
3-(3-Bromophenyl)-4H-1,2,4-triazole

Precaución: for Research Use Only

Formula: C₈H₆BrN₃
MW: 224.06 g/mol
MDL Number: MFCD18087726
CAS Number: 342617-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR