1-Amino-2,4-dibromo-9,10-anthraquinone 98%

Supplier: Ambeed
Warning

Synonyms: 1-Amino-2,4-dibromoanthraquinone

A111732-100G A111732-25G A111732-5G
76869-766EA 232.34 USD
76869-766 76869-768 76869-770
1-Amino-2,4-dibromo-9,10-anthraquinone 98%
1-Amino-2,4-dibromo-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₁₄H₇Br₂NO₂
MW: 381.03 g/mol
MDL Number: MFCD00019155
CAS Number: 81-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR