2-Bromo-4-fluoro-5-nitrotoluene 95%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-5-fluoro-2-methyl-4-nitrobenzene

A111632-10G A111632-1G A111632-250MG A111632-5G
76869-752EA 50 USD
76869-752 76869-754 76869-756 76869-758
2-Bromo-4-fluoro-5-nitrotoluene 95%
2-Bromo-4-fluoro-5-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrFNO₂
MDL Number: MFCD11847636
CAS Number: 64695-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR