(3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine hemi((2R,3R)-2,3-bis((4-methylbenzoyl)oxy)succinate) 97%

Supplier: Ambeed
Warning

Synonyms: (3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine hemi((2R,3R)-2,3-bis((4-methylbenzoyl)oxy)succinate)

A111487-10G A111487-1G A111487-25G A111487-5G
76869-740EA 51 USD
76869-740 76869-742 76869-744 76869-746
(3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine hemi((2R,3R)-2,3-bis((4-methylbenzoyl)oxy)succinate) 97%
(3R,4R)-1-Benzyl-N,4-dimethylpiperidin-3-amine hemi((2R,3R)-2,3-bis((4-methylbenzoyl)oxy)succinate)

Precaución: for Research Use Only

Formula: C₄₈H₆₂N₄O₈
MDL Number: MFCD22495378
CAS Number: 477600-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR