Benzyl (1-(hydroxymethyl)cyclopropyl)carbamate 97%

Supplier: Ambeed
Warning

A111304-1G A111304-250MG
76869-720EA 55.17 USD
76869-720 76869-722
Benzyl (1-(hydroxymethyl)cyclopropyl)carbamate 97%
Benzyl (1-(hydroxymethyl)cyclopropyl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃
MW: 221.25 g/mol
MDL Number: MFCD16294221
CAS Number: 103500-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR