2-Bromo-6-nitrobenzyl bromide 97%

Supplier: Ambeed
Danger

A110748-100MG A110748-1G A110748-250MG A110748-5G
76869-696EA 16.98 USD
76869-696 76869-698 76869-700 76869-702
2-Bromo-6-nitrobenzyl bromide 97%
2-Bromo-6-nitrobenzyl bromide

Precaución: for Research Use Only

Formula: C₇H₅Br₂NO₂
MDL Number: MFCD11036286
CAS Number: 58579-54-7
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR